Learn More
Medchemexpress LLC Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD | 1037589-69-7 | 99.8% | 528.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113697
Encompass_Preferred
Mal-amido-PEG2-C2-amido-Ph-C2-CO-AZD (Mal-PEG2-AZD) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is for research use only.
  • Used in the synthesis of PROTACs.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • For research use only.
Catalog No. 50-004-38449