Learn More
Medchemexpress LLC Mal-amido-PEG2-NHS ester | 955094-26-5 | MFCD11041137 | 99.3%
SDP
Supplier:  Medchemexpress LLC HYW04028925MG
Encompass_Preferred
Mal-amido-PEG2-NHS ester is a nonclaevable ADC linker containing a maleimide group and an NHS ester The NHS ester can be used to label the primary amines (-NH2) of proteins amine-modified oligonucleotides and other amine-containing molecules[1][2]
Catalog No. 50-002-03316