Learn More
Medchemexpress LLC Mal-amido-PEG2-TFP ester | 1431291-44-9 | 98.7% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130232
Encompass_Preferred
Mal-amido-PEG2-TFP ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This product is for research use only.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-43027