Learn More
Medchemexpress LLC Mal-Amido-PEG4-Boc | 1415800-35-9 | 99.8% | C22H36N2O9 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140967
Encompass_Preferred
Mal-Amido-PEG4-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the protein of interest. This mechanism exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52762