Learn More
Medchemexpress LLC Mal-amido-PEG4-NHS ester | 756525-99-2 | MFCD11041113 | >98.0% | 513.50 | C22H31N3O11 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW040134
Encompass_Preferred
Mal-amido-PEG4-NHS ester is a polyethylene glycol (PEG)-based bifunctional linker combining a maleimide group and an N-hydroxysuccinimide (NHS) ester. It is used in PROTAC synthesis and general bioconjugation to enable orthogonal coupling strategies: the maleimide reacts with thiols, while the NHS ester couples to amines. The short PEG4 spacer improves aqueous solubility and helps minimize steric hindrance during conjugation.
  • Bifunctional maleimide and NHS ester for orthogonal conjugation
  • PEG4 spacer increases solubility in aqueous media
  • Enables thiol-selective and amine-selective coupling reactions
  • Well-suited for PROTAC linker assembly and bioconjugation workflows
  • Supplied with analytical documentation for research use
Catalog No. 50-003-03514