Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-amido-PEG5-C2-NHS ester | 1315355-92-0 | ≥98.0% | 557.55 g·mol⁻1 | C24H35N3O12 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY126508100MG

Encompass_Preferred

Mal-amido-PEG5-C2-NHS ester is a noncleavable linker reagent combining a maleimide and an N-hydroxysuccinimide (NHS) ester for thiol- and amine-directed bioconjugation. It is intended for labeling primary amines and for constructing stable linkages in antibody-drug conjugates, protein, and oligonucleotide conjugations.

  • Contains maleimide for thiol-selective conjugation
  • Contains NHS ester reactive toward primary amines
  • High purity (≥98.0%) suitable for research use
  • Molecular weight ~557.55 and formula C24H35N3O12
  • Available in a 100 mg package for lab-scale synthesis and labeling workflows

Catalog No. 50-002-03059


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.