Learn More
Medchemexpress LLC Mal-amido-PEG6-acid | 1334177-79-5 | 99.3% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1303991G
Encompass_Preferred
Mal-amido-PEG6-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-40223