Learn More
Medchemexpress LLC Mal-amido-PEG6-acid | 1334177-79-5 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130399100MG
Encompass_Preferred
Mal-amido-PEG6-acid is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, making them suitable for research applications.
  • PEG-based PROTAC linker
  • Facilitates the synthesis of PROTACs
  • Utilizes the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • High purity (99.3%)
  • Appears as an off-white to gray solid
Catalog No. 50-003-38149