Learn More
Medchemexpress LLC Mal-amido-PEG6-NHS ester | 1137109-21-7 | 98.8% | 601.60 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW008879500MG
Encompass_Preferred
Mal-amido-PEG6-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-51202