Learn More
Medchemexpress LLC Mal-amido-PEG8-C2-acid | 1334177-86-4 | MFCD13184956 | 98.7% | 592.63 g·mol-1 | C26H44N2O13 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY101159250MG
Encompass_Preferred
Mal-amido-PEG8-C2-acid is a maleimide-functionalized PEG8 linker designed for stable, non-cleavable conjugation of payloads to thiol-containing biomolecules. It provides a hydrophilic spacer to improve solubility and reduce aggregation during antibody-drug conjugate and bioconjugation workflows.
  • Maleimide group for selective thiol conjugation.
  • PEG8 spacer to improve solubility and reduce aggregation.
  • Non-cleavable linker to provide stable payload attachment.
  • High purity suitable for research and development use (≈98.7%).
  • Solid form for straightforward handling and storage.
Catalog No. 50-002-12113