Learn More
Medchemexpress LLC Mal-amido-PEG8-NHS ester | 756525-93-6 | MFCD11041144 | 98.0% | 689.71 | C30H47N3O15 | 100 MG

Supplier: Medchemexpress LLC HYW040133100MG
Mal-amido-PEG8-NHS ester is a heterobifunctional PEG-based linker bearing a maleimide and an N-hydroxysuccinimide (NHS) ester. It is designed for efficient bioconjugation of thiol- and amine-containing molecules and is commonly used in PROTAC synthesis and other linker-driven conjugation chemistries. The PEG8 spacer improves aqueous solubility and provides a flexible linker length.
- Heterobifunctional maleimide and NHS ester for thiol and amine coupling.
- PEG8 spacer increases solubility and provides flexible linkage.
- Suitable for PROTAC construction and protein or peptide conjugation.
- High reported purity on product page.
- Soluble in DMSO (≥100 mg/mL) for in vitro use.
- Storage recommendations provided for solid and solution stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.