Learn More
Medchemexpress LLC Mal-amido-PEG8-TFP ester | 1924596-31-5 | C32H44F4N2O13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140979100MG
Encompass_Preferred
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker: one ligand binds to an E3 ubiquitin ligase, and the other binds to the target protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-54880