Learn More
Medchemexpress LLC Mal-amido-PEG8-TFP ester | 1924596-31-5 | C32H44F4N2O13 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14097950MG
Encompass_Preferred
Mal-amido-PEG8-TFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules composed of two different ligands connected by a linker, where one ligand binds to an E3 ubiquitin ligase and the other to a target protein. These molecules leverage the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Composed of two different ligands connected by a linker
  • Binds to an E3 ubiquitin ligase and a target protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Achieves selective degradation of target proteins
Catalog No. 50-003-54885