Learn More
Medchemexpress LLC Mal-amido-PEG9-acid | 2112731-43-6 | 99.5% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130856
Encompass_Preferred
Mal-amido-PEG9-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only and has not been fully validated for medical applications.
  • PEG-based linker
  • Used in synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Utilizes ubiquitin-proteasome system
  • Contains two ligands connected by a linker
Catalog No. 50-003-42668