Learn More
Medchemexpress LLC Mal-C2-gly3-eda-pnu-159682 | 2259318-53-9 | 93.4% | 991.95 | C46H53N7O18 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1266881MG
Encompass_Preferred
Mal-C2-Gly3-EDA-PNU-159682 is a warhead-linker conjugate used for antibody-drug conjugate (ADC) research, consisting of a cleavable maleimide-based Gly3-EDA linker conjugated to the potent cytotoxin PNU-159682. Supplied as a solid for research use only.
  • Warhead-linker conjugate for ADC synthesis and evaluation.
  • Cleavable maleimide-based Gly3-EDA linker enabling payload release.
  • Contains potent PNU-159682 cytotoxin for payload activity studies.
  • Supplied as a solid with reported purity suitable for research use.
  • Provided in small-format quantities for preclinical experiments.
Catalog No. 50-002-02077