Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-C6-α-amanitin | 1578249-76-9 | 98.5% | 1211.34 | C55H78N12O17S | 2 MG
SDP

Supplier:  Medchemexpress LLC HY1266832MG

Encompass_Preferred

Mal-C6-α-amanitin is an antibody-drug conjugate (ADC) payload consisting of α-amanitin covalently linked via a maleimide C6 (Mal-C6) linker. It is supplied as a solid for research use and acts as an RNA polymerase II inhibitor to provide targeted cytotoxicity.

  • Drug-linker conjugate for ADC research and payload development.
  • High purity: 98.5%.
  • Molecular weight 1211.34 Da.
  • Molecular formula C55H78N12O17S.
  • White to light yellow solid appearance.
  • Store at -20°C; in solvent, store at -80°C up to 6 months (protect from light).

Catalog No. 50-002-01965


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.