Learn More
Medchemexpress LLC Mal-C6-α-amanitin | 1578249-76-9 | 98.5% | 1211.34 | C55H78N12O17S | 5 MG

Supplier: Medchemexpress LLC HY1266835MG
Mal-C6-α-amanitin is an agent-linker conjugate for antibody-drug conjugates that delivers the RNA polymerase II inhibitor α-amanitin via a Mal-C6 linker. It is supplied as a solid for research use and shows potent antitumor activity in preclinical studies.
- Agent-linker conjugate for antibody-drug conjugates
- Molecular action: RNA polymerase II inhibitor
- Form: solid; color white to light yellow
- Purity: 98.5%
- Pack size: 5 mg
- Storage: -20°C, protect from light; in solvent -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.