Learn More
Medchemexpress LLC Mal-C6-α-amanitin | 1578249-76-9 | 98.5% | 1211.34 g/mol | C55H78N12O17S | 100 UG

Supplier: Medchemexpress LLC HY126683100UG
Mal-C6-α-amanitin is an antibody-drug conjugate (ADC) cytotoxin/linker conjugate composed of α-amanitin linked via a Mal-C6 maleimide linker. It acts as an RNA polymerase II inhibitor and is supplied as a solid for research applications, including ADC development and antitumor studies.
- Uses α-amanitin as an RNA polymerase II inhibitor.
- Supplied as a solid, white to light yellow.
- Purity 98.5%.
- Molecular weight 1211.34 g/mol.
- Formula C55H78N12O17S.
- Storage: protect from light; in solvent, store at -80°C for up to 6 months, or -20°C for up to 1 month.
- For research use only; not intended for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.