Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-C6-amine (TFA) | 731862-92-3 | MFCD11519191 | 99.7% | 310.27 g/mol | C12H17F3N2O4 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0182911G

Encompass_Preferred

Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker supplied as the trifluoroacetate salt. It contains maleimide and aminohexyl functionalities designed for use in PROTAC and bioconjugate synthesis, providing compatibility with common coupling chemistries and formulation protocols for research applications.

  • Alkyl chain linker suitable for PROTAC synthesis.
  • Maleimide and amine functional groups for versatile conjugation.
  • High reported purity (>99%) for reproducible results.
  • Available in multiple pack sizes, including 1 g.
  • Soluble in DMSO; in vivo formulation protocols provided.
  • Storage and stability guidance provided for solid and solution forms.

Catalog No. 50-003-01440


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.