Learn More
Medchemexpress LLC Mal-(CH2)5-val-cit-pab-eribulin | 2130869-21-3 | 99.8% | 1328.54 g/mol | C69H97N7O19 | 100 MG

Supplier: Medchemexpress LLC HY139642100MG
Mal-(CH2)5-Val-Cit-PAB-Eribulin is an antibody-drug conjugate (ADC) linker-payload conjugate consisting of the microtubule inhibitor eribulin covalently linked via a cleavable Mal-(CH2)5-Val-Cit-PAB peptide linker. It is supplied as a solid research reagent intended for preclinical ADC design and antitumor evaluation.
- Solid, white to yellow material for laboratory handling.
- High purity suitable for research (99.82% reported).
- Molecular weight 1,328.54 g/mol and formula C69H97N7O19.
- Available in common research quantities, including 100 MG.
- Contains eribulin as the cytotoxic payload for ADC studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.