Learn More
Medchemexpress LLC HY-100567 10mg , MAL-di-EG-Val-Cit-PAB-MMAE CAS: Purity:98%
SDP
Supplier:  Medchemexpress LLC HY10056710MG
Encompass_Preferred
Medchemexpress, HY-100567 10mg MAL-di-EG-Val-Cit-PAB-MMAE CAS: Formula:C72H112N12O18 Purity:98% MAL-di-EG-Val-Cit-PAB-MMAE consists the ADCs linker (MAL-di-EG-Val-Cit-PAB) and potent tubulin inhibitor (MMAE), MAL-di-EG-Val-Cit-PAB-MMAE is an antibody drug conjugate. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-4042