Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-di-EG-Val-Cit-PAB-MMAF | 96.0% | 1447.71 | C72H110N12O19 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY12871150MG

Encompass_Preferred

MAL-di-EG-Val-Cit-PAB-MMAF is a drug-linker conjugate used in antibody-drug conjugate research that couples a cleavable valine-citrulline p-aminobenzyl linker to monomethyl auristatin F (MMAF).

  • Drug-linker conjugate with MMAF payload.
  • Molecular formula: C72H110N12O19.
  • Molecular weight: 1447.71.
  • Purity: 95.99% initial; select batches up to 98.56%.
  • Appearance: white to off-white solid.
  • Storage: 4°C under nitrogen; unstable in solution, prepare fresh solutions.
  • Available sizes: 5 mg, 10 mg, 25 mg, 50 mg, 100 mg; larger sizes by quote.

Catalog No. 50-002-04189


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.