Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-EGGGG-PEG8-amide-bis(deoxyglucitol) | 2360920-01-8 | C55H94N10O31 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY486925MG

Encompass_Preferred

Mal-EGGGG-PEG8-amide-bis(deoxyglucitol) is a cleavable ADC linker. This product is for research use only and has not been fully validated for medical applications.

  • Target: ADC linker
  • Pathway: Antibody-drug conjugate/ADC related
  • Storage at -20°C, protected from light, and under nitrogen
  • In solvent storage at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen
  • Soluble in H₂O at 100 mg/mL (71.87 mM) with ultrasonic assistance

Catalog No. 50-003-57949


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.