Learn More
Medchemexpress LLC Mal-ggfg-pab-mmae | 3055607-18-3 | 100.0% | 1336.57 | C69H97N11O16 | 25 MG

Supplier: Medchemexpress LLC HY17051225MG
Mal-GGFG-PAB-MMAE is a drug-linker conjugate composed of the mitotic inhibitor monomethyl auristatin E (MMAE) and a maleimide-containing GGFG-PAB linker for use in antibody-drug conjugate (ADC) synthesis and payload delivery research.
- Used for antibody-drug conjugate synthesis and linker development.
- Contains MMAE payload for tubulin inhibition-based cytotoxicity.
- High measured purity suitable for research applications.
- Solid, white to off-white appearance; supplied as a 25 mg pack.
- Storage: sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.