Learn More
Medchemexpress LLC Mal-NH-PEG8-CH2CH2COOPFP ester | 2055023-14-6 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1308635G
Encompass_Preferred
Mal-NH-PEG8-CH2CH2COOPFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-40277