Learn More
Medchemexpress LLC Mal-PEG-mal (MW 2000) | 98.4% | 50 MG

Supplier: Medchemexpress LLC HY14071950MG
Mal-PEG-mal (MW 2000) is a solid chemical compound primarily used as a PROTAC linker for laboratory applications and in the manufacturing of substances.
- Used as a PROTAC linker
- Suitable for laboratory chemical applications
- Suitable for substance manufacturing
- Solid appearance
- Store at -20°C, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.