Learn More
Medchemexpress LLC Mal-PEG1-acid | 760952-64-5 | 99.9% | 213.19 g·mol⁻1 | C9H11NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126960100MG
Encompass_Preferred
Mal-PEG1-acid is a PEG-based, non-cleavable 1-unit linker used in the synthesis of antibody-drug conjugates (ADCs) and proteolysis-targeting chimeras (PROTACs). It is supplied at high purity and is soluble in DMSO, making it suitable for small-scale synthesis and linker design.
  • Non-cleavable 1-unit PEG linker suitable for ADC and PROTAC synthesis.
  • High purity to support consistent synthetic outcomes.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Available in multiple sizes for small- to medium-scale synthesis.
  • Stable in solvent: -80°C up to 6 months, -20°C up to 1 month (protect from light).
Catalog No. 50-002-03129