Learn More
Medchemexpress LLC Mal-PEG10-NHS ester | 3055858-73-3 | 98.0% | 706.73 | 25 MG
Supplier: Medchemexpress LLC 25MGHY138726
Mal-PEG10-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
- PEG-based PROTAC linker
- Used in the synthesis of PROTACs
- Exploits the intracellular ubiquitin-proteasome system
- Selectively degrades target proteins