Learn More
Cayman Chemical mal-PEG2-AZD 25mg
SDP
Supplier:  Cayman Chemical 2012925 MG
Encompass_Preferred
A bioconjugation linker; has been used in the development of protein- and peptidomimetic-antibody conjugates
Catalog No. 50-298-2603