Learn More
Medchemexpress LLC Mal-PEG2-NH2 TFA | 660843-23-2 | 99.37% | 342.27 | 100 MG

Supplier: Medchemexpress LLC HY35261A100MG
Mal-PEG2-NH2 (TFA) is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two different ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
- Appearance: oil
- Color: light yellow to yellow
- Shipping: room temperature in continental US
- Storage: -20°C, stored under nitrogen, away from moisture
- Solubility (in vitro): DMSO: 100 mg/mL (292.17 mM; requires ultrasonic)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.