Learn More
Medchemexpress LLC Mal-PEG2-Val-Cit-PAB-OH | 2055041-38-6 | 99.5% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130222100MG
Encompass_Preferred
Mal-PEG2-Val-Cit-PAB-OH is a cleavable linker used in the synthesis of antibody-drug conjugates (ADCs) and PROTACs. This linker is designed for high purity applications in research and development.
  • Cleavable linker for antibody-drug conjugates
  • Used in the synthesis of PROTACs
  • Suitable for protease cleavable applications
  • High purity: 99.5%
  • Stored under nitrogen at -20°C for optimal stability
Catalog No. 50-003-41208