Learn More
Medchemexpress LLC Mal-PEG2-Val-Cit-PABA-PNP | 1345681-52-8 | 96.8% | 755.73 | C34H41N7O13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1311565MG
Encompass_Preferred
Mal-PEG2-Val-Cit-PABA-PNP is a cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a maleimide reactive group, a two-unit polyethylene glycol spacer, and a protease-sensitive Val-Cit-PABA peptide spacer that enables payload release in target cells.
  • cleavable linker for ADC synthesis
  • contains maleimide (Mal) reactive group
  • two-unit PEG spacer (PEG2) provides solubility and flexibility
  • val-cit-paba sequence is protease sensitive for intracellular release
  • para-nitrophenyl (PNP) activating group for conjugation chemistry
  • solid, white to off-white appearance; high purity
  • available in multiple sizes and requires protection from light and storage under inert gas
Catalog No. 50-002-02024