Learn More
Medchemexpress LLC Mal-PEG3-Boc | 518044-35-4 | 97% | 357.40 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY135169
Encompass_Preferred
Mal-PEG3-Boc is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs (Proteolysis Targeting Chimeras). PROTACs function by exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based linker
  • Facilitates synthesis of PROTACs
  • Enables targeted protein degradation
  • Utilizes the ubiquitin-proteasome system
Catalog No. 50-003-66173