Learn More
Medchemexpress LLC Mal-PEG4-C2-NH2 (TFA) | 2512227-13-1 | 98.6% | 430.37 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138380100MG
Encompass_Preferred
Mal-PEG4-C2-NH2 TFA is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These compounds utilize the ubiquitin-proteasome system to selectively degrade target proteins, making them valuable laboratory chemicals for substance manufacturing.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50462