Learn More
Medchemexpress LLC Mal-PEG4-NHS ester | 1325208-25-0 | 99.4% | 442.42 | C19H26N2O10 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1265055G
Encompass_Preferred
Mal-PEG4-NHS ester is a heterobifunctional polyethylene glycol (PEG) linker bearing a terminal maleimide and an N-hydroxysuccinimide (NHS) ester. It is used to conjugate sulfhydryl-containing molecules to primary amines for bioconjugation applications such as antibody-drug conjugates and linking quantum dots to PEGylated liposomes.
  • heterobifunctional linker enabling thiol-to-amine conjugation
  • contains a four-unit peg spacer to improve solubility and reduce steric hindrance
  • maleimide group reacts selectively with sulfhydryl groups
  • nhs ester reacts with primary amines under mild conditions
  • provided as a high-purity reagent suitable for research use
Catalog No. 50-002-02446