Learn More
Medchemexpress LLC Mal-PEG4-NHS ester | 1325208-25-0 | 99.4% | 442.42 g·mol⁻1 | C19H26N2O10 | 1 G

Supplier: Medchemexpress LLC HY1265051G
Mal-PEG4-NHS ester is a PEG-based bifunctional linker containing a maleimide group and an N-hydroxysuccinimide (NHS) ester that enables conjugation between thiol-containing and amine-containing molecules. It is commonly used in antibody-drug conjugate (ADC) linker chemistry, quantum dot conjugation, and linking PEGylated liposomes.
- Non-cleavable maleimide-NHS ester linker suitable for thiol-to-amine conjugation.
- Molecular weight approximately 442.42 g·mol⁻1.
- Chemical formula C19H26N2O10.
- High purity (≈99.4%) as supplied.
- Soluble in DMSO (100 mg/mL); hygroscopic-use freshly opened DMSO and sonication as needed.
- Storage: -20°C for solid stock (up to 3 years); in solvent: -80°C (6 months), -20°C (1 month).
- Available in laboratory-scale quantities including 1 g packaging.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.