Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG4-PFP ester | 1415800-42-8 | 95.4% | 511.39 g·mol-1 | C21H22F5NO8 | 1 G
SDP

Supplier:  Medchemexpress LLC HY1265061G

Encompass_Preferred

Mal-PEG4-PFP ester is a noncleavable ADC linker that incorporates a maleimide reactive group, a four-unit polyethylene glycol (PEG4) spacer, and a pentafluorophenyl (PFP) activated ester. It is supplied for research use to enable conjugation of payloads to thiol- or amine-containing biomolecules.

  • Noncleavable ADC linker with a maleimide reactive group.
  • Four-unit PEG spacer for increased solubility and flexibility.
  • PFP ester activated for efficient coupling to amines.
  • Molecular weight 511.39 g·mol-1.
  • Chemical formula C21H22F5NO8.
  • Purity 95.4% (as supplied).
  • Storage: pure form -20°C for 3 years, 4°C for 2 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-03748


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.