Learn More
Medchemexpress LLC Mal-PEG4-val-cit-pab | 1949793-41-2 | 98.9% | 777.86 | C36H55N7O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12667225MG
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB is a protease-cleavable antibody-drug conjugate (ADC) linker containing a maleimide reactive group and a PEG4 spacer. It is intended for use in the synthesis of ADCs to attach payloads to antibodies via a Val-Cit-PAB cleavable sequence.
  • Cleavable Val-Cit-PAB dipeptide is protease sensitive.
  • Maleimide reactive group enables thiol conjugation.
  • PEG4 spacer improves solubility and reduces aggregation.
  • Molecular weight 777.86 and formula C36H55N7O12.
  • High purity typically near 98.9% and provided as a solid.
Catalog No. 50-002-01923