Learn More
Medchemexpress LLC Mal-PEG4-Val-Cit-PAB-PNP | 2112738-09-5 | MFCD30828694 | >98.0% | 871.89 g/mol | C40H53N7O15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14014510MG
Encompass_Preferred
Mal-PEG4-Val-Cit-PAB-PNP is a cleavable polyethylene glycol (PEG4) antibody-drug conjugate (ADC) linker containing a maleimide thiol-reactive group, a valine-citrulline protease-cleavable dipeptide, and a para-aminobenzyl self-immolative spacer with a p-nitrophenyl leaving group. It is used in research to attach cytotoxic payloads to antibodies via thiol conjugation and enable intracellular payload release after protease-mediated cleavage.
  • Cleavable val-cit dipeptide for protease-triggered payload release.
  • Maleimide group for thiol-selective conjugation to cysteine residues.
  • PEG4 spacer improves hydrophilicity and solubility.
  • PAB self-immolative spacer enables rapid payload liberation after cleavage.
  • Available in small pack sizes suitable for research-scale synthesis.
Catalog No. 50-002-02887