Learn More
Medchemexpress LLC Mal-PEG4-VC-PAB-DMEA-PNU-159682 | 2259318-52-8 | 1559.62 | C74H98N10O27 | 5 MG

Supplier: Medchemexpress LLC HY1266875MG
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a cleavable antibody-drug conjugate (ADC) drug-linker conjugate that couples a maleimide-PEG4 linker to the cytotoxic payload DMEA-PNU-159682 for use in ADC research and synthesis. It is supplied in small milligram pack sizes for preclinical research applications.
- Cleavable maleimide-PEG4 linker enabling thiol conjugation.
- Valine-citrulline (vc) dipeptide linker with PABC self-immolative spacer.
- Includes DMEA-PNU-159682 cytotoxic payload for ADC development.
- Supplied as a small-scale mg quantity suitable for research use.
- Molecular weight 1559.62 and molecular formula C74H98N10O27.
- CAS number 2259318-52-8 for unambiguous identification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.