Learn More
Medchemexpress LLC Mal-PEG4-vc-pab-dmea-pnu-159682 | 2259318-52-8 | 96.2% | 1559.62 | C74H98N10O27 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY12668725MG
Encompass_Preferred
Mal-PEG4-VC-PAB-DMEA-PNU-159682 is a cleavable drug-linker conjugate used for antibody-drug conjugate (ADC) research. It couples a maleimide-terminated PEG4 valine-citrulline (VC) self-immolative PAB linker to a potent cytotoxic payload to enable antibody conjugation and controlled release of the payload in target cells.
  • Cleavable maleimide-PEG4 val-cit self-immolative linker enabling intracellular release.
  • Includes a reactive maleimide moiety for thiol conjugation to antibodies.
  • Supplied as a research-grade drug-linker conjugate for ADC development workflows.
  • Provided with batch-specific certificate of analysis reporting HPLC purity and assay results.
  • Soluble in common organic solvents such as DMSO for conjugation reactions.
Catalog No. 50-002-03967