Learn More
Medchemexpress LLC Mal-PEG5-acid | 1286755-26-7 | ≥97.0% | 100 MG

Supplier: Medchemexpress LLC HY126962100MG
Mal-PEG5-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. These molecules function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. Its molecular weight is 389.40, with a chemical formula of C17H27NO9. It appears as a liquid, light brown to yellow in color, with a density of 1.249±0.06 g/cm3.
- For research use only
- Should be shipped at room temperature in the continental US
- Storage at -20°C, protected from light
- In solvent, can be stored at -80°C for 6 months or -20°C for 1 month, protected from light
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.