Learn More
Medchemexpress LLC Mal-PEG6-PFP ester | 1599486-33-5 | 96.2% | 25 MG

Supplier: Medchemexpress LLC 25MGHY130454
Mal-PEG6-PFP ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These molecules facilitate the selective degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system, utilizing two different ligands connected by this linker. One ligand binds to an E3 ubiquitin ligase, while the other targets the protein for degradation.
- PEG-based PROTAC linker
- Utilized in the synthesis of PROTACs
- Exploits the ubiquitin-proteeasome system
- Designed for selective degradation of target proteins
- Purity of 96.18%
- Store at -20°C under nitrogen
- In solvent, store at -80°C for 6 months, or -20°C for 1 month, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.