Learn More
Medchemexpress LLC Mal-PEG8-acid | 1818294-46-0 | 99.0% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130334
Encompass_Preferred
Mal-PEG8-acid is a PEG-based PROTAC linker designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins, offering a novel approach in cancer targeted therapy.
  • PEG-based PROTAC linker
  • Suitable for PROTAC synthesis
  • For research applications
Catalog No. 50-003-43029