Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mal-PEG8-acid | 1818294-46-0 | 99.04% | 1 G
SDP

Supplier:  Medchemexpress LLC HY1303341G

Encompass_Preferred

Mal-PEG8-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product has a high purity of 99.04% and is identified by CAS number 1818294-46-0. It is intended for research use only.

  • Used in PROTAC synthesis
  • PEG-based linker
  • Liquid appearance
  • Store at -20°C, protected from light
  • Shipped at room temperature in continental US

Catalog No. 50-003-39104


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.