Learn More
Medchemexpress LLC Mal-PEG8-alcohol | 2353409-63-7 | 97.1% | C20H35NO10 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1409701G
Encompass_Preferred
Mal-PEG8-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is for research use only.
  • Peg-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55127