Learn More
Medchemexpress LLC Mal-PEG8-alcohol | 2353409-63-7 | 97.1% | C20H35NO10 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140970100MG
Encompass_Preferred
Mal-PEG8-alcohol is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs exploit the ubiquitin-proteasome system
  • PROTACs selectively degrade target proteins
Catalog No. 50-003-55130