Learn More
Medchemexpress LLC Mal-PEG8-Val-Ala-PAB-Exatecan | 2679821-39-5 | 99.5% | 1329.42 | C66H85FN8O20 | 10 MG

Supplier: Medchemexpress LLC HY14727110MG
Mal-PEG8-Val-Ala-PAB-Exatecan is an antibody-drug conjugate (ADC) linker that combines a maleimide thiol-reactive group, an eight-unit polyethylene glycol (PEG8) spacer, and a cathepsin-cleavable Val-Ala-p-aminobenzyl (PAB) dipeptide to enable controlled intracellular release of payloads such as exatecan.
- Maleimide thiol-reactive group for conjugation to cysteine residues.
- Peg8 spacer reduces steric hindrance and improves solubility.
- Val-Ala-PAB dipeptide is cleavable by lysosomal proteases.
- High purity appropriate for research applications.
- Molecular weight approximately 1329.42 g/mol.
- Chemical formula C66H85FN8O20.
- Available in small-mg quantities for laboratory use (5 mg-500 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.