Learn More
Medchemexpress LLC Mal-PEG8-Val-Ala-PAB-Exatecan | 2679821-39-5 | 99.5% | 1329.42 | C66H85FN8O20 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14727150MG
Encompass_Preferred
Mal-PEG8-Val-Ala-PAB-Exatecan is an antibody-drug conjugate (ADC) linker that combines a maleimide functional group, an eight-unit polyethylene glycol (PEG8) spacer, and a cleavable Val-Ala-PAB peptide linker to connect Exatecan payloads to antibodies for targeted cancer research applications.
  • Maleimide functional group for thiol conjugation.
  • PEG8 spacer provides solubility and flexibility.
  • Cleavable Val-Ala-PAB linker enables intracellular payload release.
  • Suitable for antibody-drug conjugate synthesis and cancer research.
  • High purity (99.48%).
Catalog No. 50-002-03592