Learn More
Medchemexpress LLC Mal-PEG8-Val-Ala-PABC | 99.1% | 867.98 g/mol | C41H65N5O15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY132252100MG
Encompass_Preferred
Mal-PEG8-Val-Ala-PABC is a cleavable Tesirine linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a maleimide reactive group, a PEG8 spacer, and a Val-Ala-PABC dipeptide that is enzymatically cleavable to release payloads in lysosomal conditions. For research use only.
  • Cleavable Val-Ala-PABC dipeptide for enzymatic payload release.
  • PEG8 spacer increases hydrophilicity and solubility.
  • Maleimide reactive group for thiol conjugation to antibodies.
  • High reported purity suitable for research applications.
  • Storage: sealed and away from moisture; in solvent -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-03698